Learn More
Invitrogen™ ERV3 Polyclonal Antibody

Description
Immunogen sequence: VCYDPKSSPG IWFEIHVGSK EGDLLNQTKV FPSGKDVVSL YFDVCQIVSM GSLFPVIFSS MEYYSSCHKN RYAHPACSTD SPVTTCWDCT TWSTNQQSLG PIMLTKIPLE PDCKTSTCNS VNLTILEPDQ PIWTTGLKAP L Highest antigen sequence indentity to the following orthologs: Mouse - 21%, Rat - 21%.
Specifications
Specifications
| Antigen | ERV3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ERV3-1 |
| Gene Accession No. | Q14264 |
| Gene Alias | endogenous retroviral sequence 3; endogenous retrovirus group 3 member 1; endogenous retrovirus group 3 member 1 Env polyprotein; endogenous retrovirus group 3, member 1; Envelope polyprotein; envR; ERV3; ERV3 envelope protein; ERV-3 envelope protein; ERV3-1; ERV3-1 envelope protein; ERVR; ERV-R; ERV-R envelope protein; HERVR; HERV-R; HERV-R envelope protein; HERV-R_7q21.2 provirus ancestral Env polyprotein; SU; Surface protein; TM; Transmembrane protein |
| Gene Symbols | ERV3-1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.