Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERV3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548577

Catalog No. PA548577


Only null left
Add to Cart

Description

Description

ERV3 Polyclonal Antibody for Western Blot, IHC (P)

Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has lost its fusogenic properties. It can inhibit cell growth through decrease expression of cyclin B1 and increased expression of p21 in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ERV3
Polyclonal
Unconjugated
ERV3-1
endogenous retroviral sequence 3; endogenous retrovirus group 3 member 1; endogenous retrovirus group 3 member 1 Env polyprotein; endogenous retrovirus group 3, member 1; Envelope polyprotein; envR; ERV3; ERV3 envelope protein; ERV-3 envelope protein; ERV3-1; ERV3-1 envelope protein; ERVR; ERV-R; ERV-R envelope protein; HERVR; HERV-R; HERV-R envelope protein; HERV-R_7q21.2 provirus ancestral Env polyprotein; SU; Surface protein; TM; Transmembrane protein
Rabbit
Antigen affinity chromatography, Protein A
RUO
2086
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q14264
ERV3-1
KLH conjugated synthetic peptide between 445-474 amino acids from the C-terminal region of human ERV3.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.