Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERVFRD-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP179613

Catalog No. NBP179613


Only null left
Add to Cart

Description

Description

ERVFRD-1 Polyclonal specifically detects ERVFRD-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ERVFRD-1
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
endogenous retrovirus group FRD member 1, endogenous retrovirus group FRD, member 1, envelope polyprotein, envFRD, ERVFRDE1, FLJ41944, GLLL6191, HERV-FRD, HERV-FRD provirus ancestral Env polyprotein, HERV-FRD_6p24.1 provirus ancestral Env polyprotein, HERV-W/FRD, MGC87585, syncytin 2, UNQ6191, UNQ6191/PRO20218
Rabbit
Affinity purified
RUO
405754
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_997465
ERVFRD-1
Synthetic peptide directed towards the N terminal of human HERV-FRD. Peptide sequence TGSPYSTNCWLCTSSSTETPGTAYPASPREWTSIEAELHISYRWDPNLKG.
100 μL
Primary
Expected identity based on immunogen sequence: Lowland gorilla: 100%; Silvery gibbon: 100%; Crab-eating macaque: 100%; Chimpanzee: 100%; Bornean orangutan: 100%; Human: 100%.
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only