Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERVFRD-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | ERVFRD-1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ERVFRD-1 Polyclonal specifically detects ERVFRD-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ERVFRD-1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
405754 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
endogenous retrovirus group FRD member 1, endogenous retrovirus group FRD, member 1, envelope polyprotein, envFRD, ERVFRDE1, FLJ41944, GLLL6191, HERV-FRD, HERV-FRD provirus ancestral Env polyprotein, HERV-FRD_6p24.1 provirus ancestral Env polyprotein, HERV-W/FRD, MGC87585, syncytin 2, UNQ6191, UNQ6191/PRO20218 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 16-250 of human ERVFRD-1 (NP_997465.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title