Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERVK-21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572312
Description
ERVK-21 Polyclonal Antibody for Western Blot
Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has lost its original fusogenic properties.
Specifications
ERVK-21 | |
Polyclonal | |
Unconjugated | |
ERVK-21 | |
c12_A; endogenous retrovirus group K member 21; Endo-Rev2; envK1; EnvK1 protein; ERVK-21; PR; Protease; Proteinase; SU; TM | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100616103 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P61565 | |
ERVK-21 | |
KLH conjugated synthetic peptide between 271-302 amino acids from the Central region of human ERVK-21. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction