Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERVK-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144569 Supplier Invitrogen™ Supplier No. PA5144569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERVK-5 Polyclonal Antibody for Western Blot, ELISA

The products of the Gag polyproteins of infectious retroviruses perform highly complex orchestrated tasks during the assembly, budding, maturation, and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA. Endogenous Gag proteins may have kept, lost or modified their original function during evolution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERVK-5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ERVK-5
Gene Accession No. Q9HDB8, Q9HDB9
Gene Alias ERVK5
Gene Symbols ERVK-5
Host Species Rabbit
Immunogen Recombinant Homo sapiens ERVK-5 protein (2-667aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60358
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.