Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERVK-7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116391
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116391 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116391 Supplier Invitrogen™ Supplier No. PA5116391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERVK-7 Polyclonal Antibody for Western Blot

Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. TM anchors the envelope heterodimer to the viral membrane through one transmembrane domain. The other hydrophobic domain, called fusion peptide, mediates fusion of the viral membrane with the target cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERVK-7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ERVK-7
Gene Accession No. P61567
Gene Alias endogenous retrovirus group K member 7; ERVK7; ERVK-7; HERV-K(III); HERV-K102; HERV-KIII; PR; Protease; Proteinase; SU; TM
Gene Symbols ERVK-7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 457-491 amino acids from human ERVK-7.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 449619
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.