Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERVV-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140338 Supplier Invitrogen™ Supplier No. PA5140338
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LALDYLLAEQ GGVCAVISKS CCIYVNNSGA IEEDIKKIYD E Highest antigen sequence indentity to the following orthologs: Rat - 39%, Mouse - 39%.

Many different human endogenous retrovirus (HERV) families are expressed in normal placental tissue at high levels, suggesting that HERVs are functionally important in reproduction. This gene is part of an HERV provirus on human chromosome 19 that has inactivating mutations in the gag and pol genes. The gene's envelope protein is expressed in the human placenta but is truncated at its C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERVV-1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERVV-1
Gene Accession No. B6SEH8
Gene Alias Endogenous Retrovirus Group V, Member 1; ENVV1; HERV-V1
Gene Symbols ERVV-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERVV-1. Recombinant protein control fragment (Product #RP-109534).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.