Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Erythrocytes Monoclonal Antibody (OX-83), FITC

Catalog No. PIMA517581
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517581 500 μg
PIMA517580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517581 Supplier Invitrogen™ Supplier No. MA517581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

anti-rat erythrocyte monoclonal antibody recognizes rat erythrocytes. This antibody has also been reported to label rat neutrophils, erythrocyte stem cells and 50% bone marrow.

Erythrocytes, commonly known as red blood cells, are the primary cellular component responsible for oxygen transport within the blood. Originating from hematopoietic stem cells in the bone marrow, these cells undergo a maturation process called erythropoiesis, which necessitates key nutrients such as iron, vitamin B12, and folate. Structurally, erythrocytes have a biconcave disc shape, enhancing their surface area for gas exchange. Beyond their oxygen-carrying role, erythrocytes possess immune functions, including antigen presentation and pathogen binding, contributing to the innate immune system. Their membrane surface proteins and intracellular substances play pivotal roles in immuno-regulation, varying significantly among different disease states.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythrocytes
Applications Flow Cytometry
Classification Monoclonal
Clone OX-83
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Alias Erythroid cell marker; Ly76; Ly-76; TER119
Host Species Mouse
Immunogen Rat erythrocytes.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.