Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Erythrocytes Monoclonal Antibody (OX-83)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517577
Description
MA5-17577 targets Erythrocytes in FACS applications and shows reactivity with Rat samples. The MA5-17577 immunogen is immunogen information is proprietary.
Erythrocytes, commonly known as red blood cells, are the primary cellular component responsible for oxygen transport within the blood. Originating from hematopoietic stem cells in the bone marrow, these cells undergo a maturation process called erythropoiesis, which necessitates key nutrients such as iron, vitamin B12, and folate. Structurally, erythrocytes have a biconcave disc shape, enhancing their surface area for gas exchange. Beyond their oxygen-carrying role, erythrocytes possess immune functions, including antigen presentation and pathogen binding, contributing to the innate immune system. Their membrane surface proteins and intracellular substances play pivotal roles in immuno-regulation, varying significantly among different disease states.
Specifications
| Erythrocytes | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| OX-83 | |
| Unconjugated | |
| Erythroid cell marker; Ly76; Ly-76; TER119 | |
| Immunogen information is proprietary. | |
| 250 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction