Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin/EPO Antibody (10F3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Rat Monoclonal Antibody

Supplier:  Novus Biologicals NBP221994AF350

Catalog No. NP221994A30


Only null left
Add to Cart

Description

Description

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for ELISA.
Specifications

Specifications

Erythropoietin/EPO
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
EPO
Mouse recombinant Erythropoietin/EPO
Primary
Mouse
IgG2
ELISA
10F3
ELISA
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Rat
0.1 mL
2056
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.