Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin/EPO Antibody (10F3), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP221994AF405
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for ELISA.Specifications
Erythropoietin/EPO | |
Monoclonal | |
Alexa Fluor 405 | |
50 mM sodium borate with 0.05% sodium azide | |
EPO | |
Mouse recombinant Erythropoietin/EPO | |
Primary | |
Mouse | |
IgG2 |
ELISA | |
10F3 | |
ELISA | |
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Rat | |
0.1 mL | |
2056 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction