Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin/EPO Antibody (10F3), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP221994H
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for ELISA.Specifications
Erythropoietin/EPO | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
EPO | |
Mouse recombinant Erythropoietin/EPO | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
10F3 | |
ELISA | |
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Rat | |
Protein G purified | |
Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia | |
2056 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction