Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin/EPO Rabbit anti-Human, Clone: EPO/3793R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30715320UG
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Erythropoietin/EPO | |
Monoclonal | |
0.2 mg/ml | |
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml | |
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Recombinant fragment (around aa 28-162) of human Erythropoietin/EPO protein (exact sequence is proprietary) | |
20 μg | |
Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia | |
2056 | |
Store at 4°C. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
EPO/3793R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction