Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin R Antibody (010), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289614G
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Erythropoietin R | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse EPOR | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
10 | |
ELISA, Sandwich ELISA Detection | |
EPO-R, erythropoietin receptor, MGC138358 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Erythropoietin R (Met 1-Pro 250; NP_000112.1). | |
0.1 mL | |
Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction | |
2057 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction