Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin R Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119388AF700
Description
Erythropoietin R Polyclonal specifically detects Erythropoietin R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Erythropoietin R | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EPO-R, erythropoietin receptor, MGC138358 | |
Rabbit | |
0.1 mL | |
2057 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50 mM sodium borate with 0.05% sodium azide | |
EPOR | |
A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235] | |
Primary | |
EpoR (A364) detects endogenous levels of EpoR protein. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction