Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESAM Antibody (110), Alexa Fluor™ 750

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290757AF750
Description
ESAM Monoclonal specifically detects ESAM in Mouse samples. It is validated for ELISA.Specifications
| ESAM | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ESAM (Q925F2; Met1-Ala251). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human ESAM | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 110 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 90952 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction