Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESAM Antibody (113), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289321B
Description
ESAM Monoclonal specifically detects ESAM in Human samples. It is validated for Flow Cytometry.Specifications
ESAM | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
90952 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
113 | |
Flow Cytometry | |
2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ESAM (NP_620411.2; Met1-Ala248). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction