Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESAM Antibody (408519), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4204I
Description
ESAM Monoclonal antibody specifically detects ESAM in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
ESAM | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
90952 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry, CyTOF | |
408519 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
Mouse myeloma cell line NS0-derived recombinant human ESAM, Gln30-Ala247, Accession # Q96AP7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction