Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547368
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547368 Supplier Thermo Scientific Supplier No. PA547368
Add to Cart
Add to Cart

ESAM Polyclonal Antibody

ESAM Polyclonal Antibody for Western Blot, IHC, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen ESAM
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ESAM
Gene Accession No. Q96AP7
Gene Alias 2310008D05Rik; endothelial cell adhesion molecule; endothelial cell-selective adhesion molecule; endothelial cell-specific adhesion molecule; Esam; Esam1; HUEL (C4orf1)-interacting protein; LP4791 protein; UNQ220/PRO246; W117m
Gene Symbols ESAM
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ESAM Gln30-Ala247.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90952
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.