Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ESAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111324
Description
Immunogen sequence: DKQGKSRGHS IKTLELNVLV PPAPPSCRLQ GVPHVGANVT LSCQSPRSKP AVQYQWDRQL PSFQTFFAPA LDVIRGSL.
ESAM is a 55-kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.
Specifications
| ESAM | |
| Polyclonal | |
| Unconjugated | |
| ESAM | |
| 2310008D05Rik; endothelial cell adhesion molecule; endothelial cell-selective adhesion molecule; endothelial cell-specific adhesion molecule; Esam; Esam1; HUEL (C4orf1)-interacting protein; LP4791 protein; UNQ220/PRO246; W117m | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90952 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96AP7 | |
| ESAM | |
| Recombinant Protein corresonding to Human ESAM. Recombinant protein control fragment (Product #RP-106323). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction