Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESAM Rabbit anti-Mouse, Clone: 110, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29075750UL
Description
ESAM Monoclonal specifically detects ESAM in Mouse samples. It is validated for ELISA.Specifications
ESAM | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
90952 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
110 | |
ELISA 1:5000-1:10000 | |
2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
Recombinant Mouse ESAM protein | |
50 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction