Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESAM Recombinant Rabbit Monoclonal Antibody (202), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543093
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543093 100 μL
PIMA543454 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543093 Supplier Invitrogen™ Supplier No. MA543093
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human ESAM

ESAM is a 55-kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESAM
Applications ELISA
Classification Recombinant Monoclonal
Clone 202
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ESAM
Gene Accession No. Q96AP7
Gene Alias 2310008D05Rik; endothelial cell adhesion molecule; endothelial cell-selective adhesion molecule; endothelial cell-specific adhesion molecule; Esam; Esam1; HUEL (C4orf1)-interacting protein; LP4791 protein; UNQ220/PRO246; W117m
Gene Symbols ESAM
Host Species Rabbit
Immunogen Recombinant Human ESAM protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90952
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.