Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESAT6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982617
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2617 Supplier Bioss Supplier No. BS13107R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mycobacterium tuberculosis is a slow-growing obligate aerobic bacillus that causes most cases of tuberculosis (TB). It is a small, rod-like microbe that can withstand weak disinfectants and survive in a dry state for weeks, but can only grow within a host organism. M. tuberculosis has a thick waxy cell wall that is responsible for the typical caseous granuloma formation in tuberculosis. ESAT6 is a protein secreted by M. tuberculosis that induces a strong immune response in infected organisms, making it a strong candidate for the development of a vaccine against tuberculosis. ESAT6 may also be useful in the detection of the disease. Deletion of the ESAT6 gene leads to the loss of virulence of M. tuberculosis.

Specifications

Antigen ESAT6
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Mycobacterium tuberculosis ESAT6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.