Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli Ribonucleic acid polymerase Rabbit anti-Bacteria, Polyclonal, Nordic
SDP

Catalog No. 502101640
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1640 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1640 Supplier Vector Laboratories Supplier No. NE188/7S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Escherichia coli Ribonucleic acid polymerase Polyclonal antibody specifically detects Escherichia coli Ribonucleic acid polymerase in Bacteria samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence

Specifications

Antigen Escherichia coli Ribonucleic acid polymerase
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH
Gene rpoA
Host Species Rabbit
Immunogen Escherichia coli Ribonucleic acid polymerase
Purification Method Protein G
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947794
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.