Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Escherichia Virus Capsid Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144462 Supplier Invitrogen™ Supplier No. PA5144462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Escherichia Virus Capsid Polyclonal Antibody for Western Blot, ELISA

Capsid protein self-assembles to form an icosahedral capsid with a T=3 symmetry, about 26 nm in diameter, and consisting of 89 capsid proteins dimers (178 capsid proteins) (PubMed:27671640, PubMed:19913556). Involved in viral genome encapsidation through the interaction between a capsid protein dimer and the multiple packaging signals present in the RNA genome (PubMed:8943226, PubMed:27671640). Binding of the capsid proteins to the viral RNA induces a conformational change required for efficient T=3 shell formation (PubMed:19913556). The capsid contains also 1 copy of the A2 maturation protein (PubMed:27671640).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Escherichia Virus Capsid
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Capsid protein
Host Species Rabbit
Immunogen Recombinant Escherichia phage Qbeta (Bacteriophage Q-beta) CAPSD protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.