Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Escitalopram oxalate
GREENER_CHOICESDP

Catalog No. 479650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-965-0 50 mg
47-961-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-965-0 Supplier R&D Systems™ Supplier No. 4796/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective serotonin reuptake inhibitor (SSRI)

Escitalopram oxalate is a selective 5-HT reuptake inhibitor (Ki values are 0.89, 8150, and 10500 nM for serotonin, noradrenalin, and dopamine transporters respectively). Escitalopram oxalate is an S-enantiomer and eutomer of citalopram (Cat. No. 1427).

Chemical Identifiers

CAS 219861-08-2
Molecular Formula C22H23FN2O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 414.43
Purity 98%
Molecular Weight (g/mol) 414.433
Chemical Name or Material Escitalopram oxalate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.