Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESCO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560000 Supplier Invitrogen™ Supplier No. PA560000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERAPENCHLA NEIKPSDPPL DNQMKHSFDS ASNKNFSQCL ESKLENSPVE NVTAASTLLS QAKIDTGENK FPGSAPQQHS ILSNQTSKSS DNRETPRNHS LPKCNSHLEI Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 64%.

EFO1 (Establishment factor-like protein 1), also known as ESCO1 (establishment of cohesion 1 homolog1) or ESO1, is a member of the acetyltransferase family (GCN5 subfamily). It is a ubiquitously expressed nuclear protein that plays an important role in sister chromatid cohesion. At its C-terminus, EFO1 contains an H2C2 zinc finger motif and an acetyltransferase domain that exhibits acetyltransferase activity in vivo. Its N-terminus, containing two domains that are similar o alpha- and beta-type linker histone proteins, is essential for EFO1 association with chromosomes. EFO1 is responsible for coupling cohesion and DNA replication processes thereby ensuring proper pairing of sister chromatids. EFO1 is phosphorylated during mitosis and this may act to regulate EFO1 activity. Due to alternative splicing events, three EFO1 isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESCO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESCO1
Gene Accession No. Q5FWF5
Gene Alias A930014I12Rik; CTF; CTF7 homolog 1; ECO1; ECO1 homolog 1; EFO1; EFO1p; ESCO1; ESO1; ESO1 homolog 1; establishment factor-like protein 1; Establishment of cohesion 1 homolog 1; establishment of cohesion 1 homolog 1 (S. cerevisiae); establishment of sister chromatid cohesion N-acetyltransferase 1; establisment of cohesion 1 homolog 1; hEFO1; Kiaa1911; N-acetyltransferase ESCO1; N-acetyltransferase ESCO1 variant 2
Gene Symbols ESCO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ESCO1. Recombinant protein control fragment (Product #RP-98281).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.