Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESCO2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10087021DL594
Description
ESCO2 Polyclonal antibody specifically detects ESCO2 in Human samples. It is validated for Western BlotSpecifications
ESCO2 | |
Polyclonal | |
Western Blot | |
2410004I17Rik, EC 2.3.1.-, ECO1 homolog 2, EFO2, Establishment of cohesion 1 homolog 2, establishment of cohesion 1 homolog 2 (S. cerevisiae), N-acetyltransferase ESCO2, RBS, Roberts syndrome | |
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human establishment of cohesion 1 homolog 2 using the numbering given in entry NP_001017420.1 (GeneID 157570). | |
0.1 mL | |
Lipid and Metabolism | |
157570 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction