Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESCO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565813 Supplier Thermo Scientific Supplier No. PA565813
Add to Cart
Add to Cart

ESCO2 Polyclonal Antibody

ESCO2 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ESCO2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESCO2
Gene Accession No. Q56NI9
Gene Alias 2410004I17Rik; D030072L07Rik; ECO1 homolog 2; EFO2; Esco2; Establishment factor-like protein 2; Establishment of cohesion 1 homolog 2; establishment of cohesion 1 homolog 2 (S. cerevisiae); establishment of sister chromatid cohesion N-acetyltransferase 2; N-acetyltransferase ESCO2; RBS
Gene Symbols ESCO2
Host Species Rabbit
Immunogen Recombinant Human ESCO2. Recombinant protein control fragment (Product #RP-106486).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.