Learn More
ESCO2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301689AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.Specifications
ESCO2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q56NI9 | |
ESCO2 | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ESCO2 | |
2410004I17Rik, EFO2, RBS, ECO1 homolog 2, N-acetyltransferase ESCO2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157570 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.