Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Esculin Sesquihydrate 98.0+%, TCI America™
SDP

Catalog No. E002425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E002425G 25 g
E00245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E002425G Supplier TCI America Supplier No. E002425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66778-17-4
Molecular Formula C30H38O21
Molecular Weight (g/mol) 734.613
MDL Number MFCD00006879
InChI Key LGMHQDJZWHRZPO-HPPUHPNBSA-N
Synonym esculin sesquihydrate
PubChem CID 131675424
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 25 g
Formula Weight 340.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Esculin Sesquihydrate
RTECSNumber DJ3085000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.