Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESD Mouse anti-Human, Clone: J0-8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP266825
Description
ESD Monoclonal specifically detects ESD in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ESD | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 | |
EC 3.1.2.12, esterase 10, esterase Desterase D/formylglutathione hydrolase, FGH, FLJ11763, S-formylglutathione hydrolase | |
Mouse | |
Immunogen affinity purified | |
Proteases & Other Enzymes | |
2098 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2c |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
J0-8 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ESD | |
Recombinant protein within Human ESD aa 1-182/ 282. (SwissProt: P10768 Human) | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction