Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586038 Supplier Invitrogen™ Supplier No. PA586038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ESD Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes a serine hydrolase that belongs to the esterase D family. The encoded enzyme is active toward numerous substrates including O-acetylated sialic acids, and it may be involved in the recycling of sialic acids. This gene is used as a genetic marker for retinoblastoma and Wilson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene ESD
Gene Accession No. P10768, Q9R0P3
Gene Alias Es10; Es-10; Esd; Esterase 10; esterase D; esterase D/formylglutathione hydrolase; FGH; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; Sid 478; sid478; testicular tissue protein Li 66
Gene Symbols ESD
Host Species Rabbit
Immunogen Recombinant protein within Human ESD aa 1-282.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13885, 2098
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.