Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115199 Supplier Invitrogen™ Supplier No. PA5115199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ESD.

This gene encodes a serine hydrolase that belongs to the esterase D family. The encoded enzyme is active toward numerous substrates including O-acetylated sialic acids, and it may be involved in the recycling of sialic acids. This gene is used as a genetic marker for retinoblastoma and Wilson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ESD
Gene Accession No. B0BNE5, P10768, Q9R0P3
Gene Alias Es10; Es-10; Esd; Esterase 10; esterase D; esterase D/formylglutathione hydrolase; FGH; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; Sid 478; sid478; testicular tissue protein Li 66
Gene Symbols ESD
Host Species Rabbit
Immunogen A synthesized peptide derived from human ESD(Accession P10768), corresponding to amino acid residues S63-P113.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13885, 2098, 290401
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.