Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESE-1 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277970
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7970 Supplier Leinco Technologies Inc Supplier No. 34005100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ESE-1 Polyclonal antibody specifically detects ESE-1 in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen ESE-1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ELF3
Gene Accession No. P78545
Gene Alias E74 like ETS transcription factor 3; E74-like factor 3; E74-like factor 3 (ets domain transcription factor); E74-like factor 3 (ets domain transcription factor, epithelial-specific ); E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific); ELF3; elf3 {ECO:0000312; EMBL:AAH97980.1}; Epithelial-restricted with serine box; epithelium-restricted Ets protein ESX; Epithelium-specific Ets transcription factor 1; EPR-1; Ert; ESE-1; ESX; ets domain transcription factor, serine box (epithelial-specific); ETS-related transcription factor Elf-3; JEN; RP11-510N19.1
Host Species Rabbit
Immunogen N-terminal sequence (1-16) MAATCEISNIFSNYFS and C-terminal sequence (357-371) SSGWKEEEVLQSRN of human ESE-1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1999
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.