Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eserine Salicylate, MP Biomedicals

Catalog No. MP521376701
Click to view available options
Quantity:
1 g

  • Important Notice: This product belongs to MP Biomedicals Rare Chemical library
  • Most Rare Chemical products are available for immediate delivery and sold 'As Is'
  • MP Biomedicals does not collect analytical data for those products
  • However, analytical test may be performed upon custom request for bulk orders

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-64-7
Molecular Formula C22H27N3O5
Molecular Weight (g/mol) 413.474
MDL Number MFCD00135659
InChI Key HZOTZTANVBDFOF-PBCQUBLHSA-N
Synonym physostigmine salicylate, unii-2046zro9vu, antilirium tn, 3ar,8bs-3,4,8b-trimethyl-2,3a-dihydro-1h-pyrrolo 2,3-b indol-7-yl n-methylcarbamate;2-hydroxybenzoic acid, physostigmine salicylate usp:jan, spectrum_000223, opera_id_194, physostigmine salicylate salt, physostigmine; salicyclic acid, spectrum1500486
PubChem CID 657348
ChEBI CHEBI:48883
IUPAC Name [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate;2-hydroxybenzoic acid
SMILES CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC)C)C.C1=CC=C(C(=C1)C(=O)O)O

Specifications

Melting Point 181°C to 183°C (literature)
Linear Formula C15H21N3O2 . C7H6O3
Quantity 1 g
Formula Weight 413.45
Chemical Name or Material ESERINE SALICYLATE
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.