Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESF1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171920AF350
Description
ESF1 Polyclonal antibody specifically detects ESF1 in Human samples. It is validated for Western BlotSpecifications
ESF1 | |
Polyclonal | |
Western Blot | |
ABT1-associated protein, ABTAP, bA526K24.1, C20orf6, chromosome 20 open reading frame 6, ESF1 homolog, ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae), FLJ20368 | |
The immunogen for this product maps to a region between residue 801 and 851 of human ABT1-Associated Protein using the numbering given in entry NP_057733.2 (GeneID 51575). | |
0.1 mL | |
Stem Cell Markers | |
51575 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction