Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561419 Supplier Invitrogen™ Supplier No. PA561419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEMALLMMDE DEDSKKHFNY NKIVEHQNLS KKKKKQLMKK KELIEDDFEV NVNDARFQAM YTSHLFNLDP SDPNFKKTKA ME Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

May constitute a novel regulatory system for basal transcription. Negatively regulates ABT1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESF1
Gene Accession No. Q9H501
Gene Alias 2610101J03Rik; ABT1-associated protein; Abtap; AW545818; bA526K24.1; C20orf6; C79684; Esf1; ESF1 homolog; ESF1 nucleolar pre-rRNA processing protein homolog; ESF1, nucleolar pre-rRNA processing protein, homolog; ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae); ESF1-like protein; ESF1-like protein-like protein; HDCMC28P; RGD1306067
Gene Symbols ESF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ESF1. Recombinant protein control fragment (Product #RP-99316).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.