Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESF1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ESF1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ESF1 Polyclonal specifically detects ESF1 in Human samples. It is validated for Western Blot.Specifications
ESF1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS buffer, 2% sucrose | |
51575 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ABT1-associated protein, ABTAP, bA526K24.1, C20orf6, chromosome 20 open reading frame 6, ESF1 homolog, ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae), FLJ20368 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human ESF1 (NP_057733). Peptide sequence QELTQAIKKKESEIEKESQRKSIDPALSMLIKSIKTKTEQFQARKKQKVK | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title