Learn More
Restek™ ESI Electrode Gauge
SDP
Catalog No. 06611890 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
For Use With (Equipment):
Sciex Turbo V Sources
Catalog No. For Use With (Equipment)
06-611-890 Sciex Turbo V Sources
1 options
Catalog No. 06-611-890 Supplier Restek™ Supplier No. 25832
For Sciex Turbo V sources
  • For installing/replacing ESI electrodes in Turbo V ion source probes
  • Designed to ensure correct electrode exposure distance from probe tip
  • Provides quick and easy visual confirmation of electrode exposure
Specifications
Type ESI Electrode Gauge
For Use With (Application) Installing/replacing ESI electrodes into source probe
For Use With (Equipment) Sciex Turbo V Sources