Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESI Tuning Mix, for Ion Trap for mass spectrometry, MilliporeSigma™ Supelco™

Catalog No. 111016589 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mL

Catalog No. 11-101-6589

Supplier: MilliporeSigma Supelco 0003610ML

Only null left
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8
Flash Point 2.0°C
Purity Grade Notes Concentration of analyte in solution is μg/mL ± 0.5%, uncertainty based upon balance and Class A volumetric glassware.
Type Tuning Mix
Quantity 10 mL
Grade MS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.