Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ESPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555941
Description
Immunogen sequence: SMPAWRRDLL RKKLEEEREQ KRKEEERQKQ EELRREKEQS EKLRTLGYDE SKLAPWQRQV ILKK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Multifunctional actin-bundling protein. Plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells.
Specifications
| ESPN | |
| Polyclonal | |
| Unconjugated | |
| Espn | |
| actin cytoskeletal regulatory protein; autosomal recessive deafness type 36 protein; DFNB36; Ectoplasmic specialization protein; espin; Espn; je; jerker; Jerker, deafness locus; LP2654 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83715 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B1AK53 | |
| Espn | |
| Recombinant protein corresponding to Human ESPN. Recombinant protein control fragment (Product #RP-94521). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction