Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ESPN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572483
Description
ESPN Polyclonal Antibody for Western Blot, IHC (P), Flow
Multifunctional actin-bundling protein. Plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells.
Specifications
ESPN | |
Polyclonal | |
Unconjugated | |
Espn | |
actin cytoskeletal regulatory protein; autosomal recessive deafness type 36 protein; DFNB36; Ectoplasmic specialization protein; espin; Espn; je; jerker; Jerker, deafness locus; LP2654 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
83715 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
B1AK53 | |
Espn | |
KLH conjugated synthetic peptide between 17-45 amino acids from the N-terminal region of human ESPN. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction