Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck ESR-Chex™ Controls
SDP

Catalog No. p-108829 Shop All Streck Laboratories Products
Click to view available options
Quantity:
2 x 9 mL
4 x 9 mL
8 x 9 mL
12 x 9 mL
Level:
1, 2

Catalog No. 23-046578

Supplier: Streck 214102

Only null left
Add to Cart
Add to Cart

ESR-Chex is a whole blood sed-rate control for automated and manual ESR testing methods. It is available in two levels and assayed for the most frequently used sedimentation rate tubes in conjunction with the Classical Westergren, Modified Westergren and Wintrobe methods.

  • Ready-to-use whole blood control that resembles patient sample.
  • Available in two levels: normal and abnormal.
  • Assayed for both manual and automated sed-rate testing methods.
  • 95-day open-vial stability, 365-day closed-vial stability.
  • Alerts the technologist to variables that may affect the accuracy of patient results.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Certifications/Compliance Internal quality control standards and regulatory compliance
Compatible Methods Both automated and manual ESR methods
Content And Storage 2°C to 10°C
For Use With (Equipment) Clinical chemistry analyzer
Type ESR-Chex Control
Intended Use Verify the precision and accuracy of both manual and automated ESR methods
Level 1, 2
Quantity 2 x 9 mL
Stability 95 Days Open, 12 Months Closed Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.