Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESR1/ER Monoclonal Antibody (5C1), Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM51124M

This item is not returnable.
View return policy
Description
ESR1/ER Monoclonal antibody specifically detects ESR1/ER in Human samples. It is validated for Western BlotSpecifications
ESR1/ER | |
Monoclonal | |
0.5 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P03372 | |
Mouse | |
Protein G | |
RUO | |
2099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
5C1 | |
Unconjugated | |
ESR1 | |
DKFZp686N23123; ER; Er alpha; ER36; ERa; ERalpha; ER-alpha; ERalpha protein; ESR; esr 1; Esr1; esr-1; ESRA; Estr; Estra; Estradiol receptor; estrogen nuclear receptor alpha; Estrogen receptor; estrogen receptor 1; estrogen receptor 1 (alpha); estrogen receptor alpha; estrogen receptor alpha c-terminus splice variant 1-2; estrogen receptor alpha E1-E2-1-2; estrogen receptor alpha E1-N2-E2-1-2; estrogen receptor alpha splice variant, CTERP-1; estrogen receptor alpha splice variant, ERalphaDup5; estrogen receptor alpha splice variant, ERalphai45a; estrogen receptor alpha splice variant, ERalphai45bL; estrogen receptor alpha splice variant, ERalphai45bS; estrogen receptor alpha splice variant, ERalphai45c; estrogen receptor alpha splice variant, ERalphai56; estrogen receptor alpha splice variant, ERalphai67; estrogen receptor alpha variant delta 4; estrogen receptor protein; estrogen receptor, alpha; ESTRR; hER-alpha36; I79_001166; Nr3a1; nuclear receptor; nuclear receptor subfamily 3 grou | |
This ESR1/ER antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 246-273 amino acids from human ESR1/ER. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction