Learn More
ESR1 Mouse anti-Canine, Human, Mouse, Clone: OTI3G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807239
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.Specifications
| ESR1 | |
| Monoclonal | |
| Unconjugated | |
| ESR1 | |
| ER, ESR, ESRA, ESTRR, Era, NR3A1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 13982, 2099, 403640 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P03372, P19785 | |
| ESR1 | |
| Human recombit protein fragment corresponding to amino acids 1-310 of human ESR1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.