Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESR2 Mouse anti-Human, Clone: ERb455 (PE), Abnova™

Catalog No. 89034310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-310 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-310 Supplier Abnova Corporation Supplier No. MAB14209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human ESR2.

  • High specificity
  • High reactivity

Specifications

Antigen ESR2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone ERb455
Conjugate PE
Description Mouse monoclonal antibody raised against partial recombinant human ESR2.
Dilution Flow Cytometry (5 uL/test/million cells in 0.1 mL or 5 uL/100 uL of whole blood) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. Q92731
Gene Alias ER-BETA/ESR-BETA/ESRB/ESTRB/Erb/NR3A2
Gene Symbols ESR2
Host Species Mouse
Immunogen Recombinant protein corresponding to the C-terminus of human ESR2.
Purification Method Protein A purification
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2100
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.