Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESR2 Mouse anti-Human, Mouse, Clone: 5D2C9, Proteintech
SDP

Catalog No. 501736300 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6300 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6300 Supplier Proteintech Group Inc Supplier No. 601971IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The human ER-beta is a newly discovered estrogen receptor initially cloned and characterized from testis. The size and structure of ER-beta is very similar to ER-alpha with the ligand and DNA binding domains being highly conserved, while the amino terminus which serves as their transactivation domain has diverged significantly. Similar in function to ER-alpha ER-beta binds to estrogen with a high affinity and regulates estrogen specific gene activation through direct interaction with estrogen response elements (ERE′s).

Specifications

Antigen ESR2
Applications Western Blot
Classification Monoclonal
Clone 5D2C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ESR2
Gene Accession No. O08537, Q92731
Gene Alias ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2
Gene Symbols ESR2
Host Species Mouse
Immunogen ESR2 Fusion Protein Ag5103
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13983, 2100
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.