Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589948 Supplier Invitrogen™ Supplier No. PA589948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTASPDYLVV LFGITAGATG AKLGSDEKEL ILLFWKVVDL ANKKVGQLHE VLVRPDQLEL TEDCKEETKI DVESLSSASQ LDQALRQFNQ SVSNELNIGV GTSFCLCTDG QLHVRQILHP EASKKNVLLP ECFYSFFDLR KEFKKCCPGS PDIDKLDVAT MTEYLNFEKS SSVSRYGASQ VEDMGNIILA MISEPYNHRF SDPERVNYKF ESGTCSKMEL; Positive Samples: LO2, MCF7, HT-29, 293T, Mouse thymus, Mouse pancreas; Cellular Location: Nucleus.

RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ESRP1
Gene Accession No. Q3US41, Q6NXG1
Gene Alias 2210008M09Rik; A630065D16; BC031468; epithelial splicing regulatory protein 1; Esrp1; Rbm35a; RGD1560481; RMB35A; RNA binding motif protein 35A; RNA-binding motif protein 35A; RNA-binding protein 35A
Gene Symbols ESRP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ESRP1 (NP_0601672).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207920, 54845
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.